Merck
  • Integrated approach for obtaining bioactive peptides from whey proteins hydrolysed using a new proteolytic lactic acid bacteria.
Integrated approach for obtaining bioactive peptides from whey proteins hydrolysed using a new proteolytic lactic acid bacteria.
Food chemistry (2020-01-07)
Paulina Worsztynowicz, Wojciech Białas, Włodzimierz Grajek
ZUSAMMENFASSUNG
In this study, we investigated the ability of Enterococcus faecalis 2/28, isolated from artisan cheese, to release biopeptides from whey proteins. We used an in silico approach for predicting the bioactivities of peptides generated by E. faecalis. The results of the in vitro study showed that the whey protein hydrolysates (WPHs) obtained had angiotensin-I-converting enzyme (ACE) and dipeptidyl peptidase IV (DPP-IV) inhibitory activities, with inhibition of ACE being stronger than that of DPP-IV. To identify peptides that may be potential inhibitors of ACE, WPH with the highest ACE inhibitory activity was analysed using Sephadex G-75 gel filtration chromatography, Superdex peptide 10/300 GL size exclusion chromatography, and liquid chromatography-electrospray ionisation tandem mass spectrometry (LC-ESI-MS/MS). Among the identified peptides were ACE-inhibitory peptides (LDAQSAPLR, LKGYGGVSLPEW, and LKALPMH), antimicrobial peptides (AASDISLLDAQSAPLR, IIAEKTKIPAVF, IDALNENK, and VLVLDTDYK), DPP-IV-inhibitory peptides (LKALPMH, LKPTPEGDLEIL, LKGYGGVSLPE, LKPTPEGDLE, ILDKVGINY, and VLVLDTDYK), proliferation stimulating peptide (IDALNENK), and cytotoxic peptide (LIVTQTMK).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Sigma-Aldrich
N-Hexanoyl-L-homoserin-lacton, ≥96% (HPLC)