Merck
  • The antipsychotic sultopride is overdosed--a PET study of drug-induced receptor occupancy in comparison with sulpiride.
The antipsychotic sultopride is overdosed--a PET study of drug-induced receptor occupancy in comparison with sulpiride.
The international journal of neuropsychopharmacology (2005-11-18)
Akihiro Takano, Tetsuya Suhara, Fumihiko Yasuno, Kazutoshi Suzuki, Hidehiko Takahashi, Takuya Morimoto, Young-Joo Lee, Hiroyuki Kusuhara, Yuichi Sugiyama, Yoshiro Okubo
ZUSAMMENFASSUNG
Conventional antipsychotics tend to elicit extrapyramidal symptoms at clinical doses, but dose optimization could reduce the risk of such side-effects. In-vivo receptor-binding studies have suggested that 70-80% of dopamine D2 receptor occupancy provides the desired antipsychotic effects without extrapyramidal symptoms. In terms of dose optimization based on the occupancy, there has not been enough supporting data regarding the clinical doses of the respective antipsychotics. In this study, we measured dopamine D2 receptor occupancy of two conventional benzamide antipsychotics, sulpiride and sultopride, using positron emission tomography, to investigate the rationale of their clinical dose. Although they are prescribed at similar doses (300-1200 mg), the doses required to obtain similar receptor occupancy (70-80%) were quite different: 1010-1730 mg for sulpiride but 20-35 mg for sultopride. In terms of dose, sultopride has about 50 times greater potency than sulpiride based on dopamine D2 receptor occupancy. Evidence for the optimal doses of conventional antipsychotics based on dopamine D2 receptor occupancy would be helpful for rational antipsychotic therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sultopride hydrochloride, ≥98% (HPLC)