Merck
  • The Chinese medicinal herb decoction QRZSLXF enhances anti-inflammatory effect in TNBS-induced colitis via balancing Th17/Tregs differentiation.
The Chinese medicinal herb decoction QRZSLXF enhances anti-inflammatory effect in TNBS-induced colitis via balancing Th17/Tregs differentiation.
Journal of ethnopharmacology (2020-01-10)
Man Zhang, Heng Fan, Songwei Tan, Qing Tang, Xingxing Liu, Dongmei Zuo, Yi Liao, Zhen Nan, Chen Tan
ZUSAMMENFASSUNG
Inflammatory bowel disease (IBD) is one of the most common chronic inflammatory illnesses of the gastrointestinal tract due to the imbalance of immune homeostasis of T helper cells and/or regulatory T cells (Tregs). The Traditional Chinese medicine herb has been clinically proven for use in the treatment of IBD but its possible mechanism remains unknown. The study aims to assess the effect of Chinese medicinal herb decoction QRZSLXF (Qing Re Zao Shi Liang Xue receipt) for the treatment of TNBS-induced experimental colitis in mice and explore its relevant mechanism involved in Th17 and Tregs. Mice colitis was induced by 50% 2,4,6-Trinitrobenzenesulfonic Acid (TNBS) ethanol solution weekly manner. These established model mice were divided into model control (0.8% NaCl treatment), FICZ, naphthoflavone (NaFTV), dexamethasone (DXM), and QRZSLXF (QrLx) groups. The colonoscopy, H&E staining, and immune staining were used to analyze the disease severity, inflammatory condition and Th17 or Treg related factors expression. High-performance liquid chromatography-mass spectrometry (HPLC/MS) was used to assess the content of FICZ in the colon tissues. Western blot and ELISA were used to examine the expression of Th17 or Treg related factors protein levels. Flow cytometry analysis was performed to assess the number and ratio of Th17/Tregs in splenocytes, and mesenteric lymph node lymphocytes (MLNCs), and lamina propria mononuclear cells (LPMCs). NaFTV, DXM and QrLx groups intestinal inflammation scores were significantly lower than that in colitis model control and FICZ groups, while the IL-6, STAT3, and RORγt expression levels were significantly lower than those in the model control and FICZ groups. Mass spectrometry results showed FICZ that in both DXM and QrLx groups was lower than control model and FICZ groups. Flow cytometry results showed that DXM, NaFTV and QrLx could significantly reduce Th17 proportion and increase Treg proportion in splenocytes, MLNCs, and LPMCs. NaFTV and QrLx treatment could decrease symptoms and inflammatory colitis, by decreasing of FICZ concentration and AhR signaling in colon, resulting in reducing the expression of IL-6, STAT3, and RORγt, whereas increasing the expression of FOXP3, consequently reducing the proportion of Th17 cells and increasing the proportion of Treg cells, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Picrylsulfonsäure -Lösung, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
α-Naphthoflavon, ≥98%