Merck
  • Biopolymer K-carrageenan wrapped ZnO nanoparticles as drug delivery vehicles for anti MRSA therapy.
Biopolymer K-carrageenan wrapped ZnO nanoparticles as drug delivery vehicles for anti MRSA therapy.
International journal of biological macromolecules (2019-12-11)
Sekar Vijayakumar, Kandasamy Saravanakumar, Balasubramanian Malaikozhundan, Mani Divya, Baskaralingam Vaseeharan, Esteban F Durán-Lara, Myeong-Hyeon Wang
ZUSAMMENFASSUNG
Kappa-Carrageenan wrapped ZnO nanoparticles (KC-ZnO NPs) was synthesized, physico-chemically characterized and evaluated their biocompatibility and antimicrobial therapy against MRSA. XRD showed the highly crystalline and hexagonal phase structure of ZnO NPs. FETEM confirmed the spherical and hexagonal shaped particle with the mean size of 97.03 ± 9.05 nm. The synthesized KC-ZnO NPs exhibited significant antibacterial activity against MRSA. The biofilm growth of MRSA was greatly inhibited at 100 μg/ml as observed through live and dead cell assay. KC-ZnO NPs have shown invitro anti-inflammatory activity (82%) at 500 μg/ml. KC-ZnO NPs was non-toxic to NIH3T3 mouse embryonic fibroblasts cell lines. Further, no apoptotic and necrotic mediated death in NIH3T3 mouse embryonic fibroblasts cells were noticed by flow cytometric analysis. KC-ZnO NPs have good biocompatibility as recorded by the least hemolysis percentage (<3%) up to 100 μg/ml, which is much lesser than the acceptable limit. In addition, ecosafety analysis has shown that KC-ZnO NPs and kappa karrageenan (0-500 μg/ml) caused no mortality of A. salina after 48 h. However, bare zinc acetate has shown 35% mortality of A. salina after 48 h. The results conclude that KC-ZnO NPs could be a novel antibacterial therapy for the treatment of MRSA associated infectious.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acridinorange -hydrochlorid -Lösung, 10 mg/mL in H2O, ≥95.0% (HPLC)