Merck
  • Secreted frizzled-related protein 2 (SFRP2) expression promotes lesion proliferation via canonical WNT signaling and indicates lesion borders in extraovarian endometriosis.
Secreted frizzled-related protein 2 (SFRP2) expression promotes lesion proliferation via canonical WNT signaling and indicates lesion borders in extraovarian endometriosis.
Human reproduction (Oxford, England) (2018-02-21)
T Heinosalo, M Gabriel, L Kallio, P Adhikari, K Huhtinen, T D Laajala, E Kaikkonen, A Mehmood, P Suvitie, H Kujari, T Aittokallio, A Perheentupa, M Poutanen
ZUSAMMENFASSUNG
What is the role of SFRP2 in endometriosis? SFRP2 acts as a canonical WNT/CTNNB1 signaling agonist in endometriosis, regulating endometriosis lesion growth and indicating endometriosis lesion borders together with CTNNB1 (also known as beta catenin). Endometriosis is a common, chronic disease that affects women of reproductive age, causing pain and infertility, and has significant economic impact on national health systems. Despite extensive research, the pathogenesis of endometriosis is poorly understood, and targeted medical treatments are lacking. WNT signaling is dysregulated in various human diseases, but its role in extraovarian endometriosis has not been fully elucidated. We evaluated the significance of WNT signaling, and especially secreted frizzled-related protein 2 (SFRP2), in extraovarian endometriosis, including peritoneal and deep lesions. The study design was based on a cohort of clinical samples collected by laparoscopy or curettage and questionnaire data from healthy controls and endometriosis patients. Global gene expression analysis in human endometrium (n = 104) and endometriosis (n = 177) specimens from 47 healthy controls and 103 endometriosis patients was followed by bioinformatics and supportive qPCR analyses. Immunohistochemistry, Western blotting, primary cell culture and siRNA knockdown approaches were used to validate the findings. Among the 220 WNT signaling and CTNNB1 target genes analysed, 184 genes showed differential expression in extraovarian endometriosis (P < 0.05) compared with endometrium tissue, including SFRP2 and CTNNB1. Menstrual cycle-dependent regulation of WNT genes observed in the endometrium was lost in endometriosis lesions, as shown by hierarchical clustering. Immunohistochemical analysis indicated that SFRP2 and CTNNB1 are novel endometriosis lesion border markers, complementing immunostaining for the known marker CD10 (also known as MME). SFRP2 and CTNNB1 localized similarly in both the epithelium and stroma of extraovarian endometriosis tissue, and interestingly, both also indicated an additional distant lesion border, suggesting that WNT signaling is altered in the endometriosis stroma beyond the primary border indicated by the known marker CD10. SFRP2 expression was positively associated with pain symptoms experienced by patients (P < 0.05), and functional loss of SFRP2 in extraovarian endometriosis primary cell cultures resulted in decreased cell proliferation (P < 0.05) associated with reduced CTNNB1 protein expression (P = 0.05). SFRP2 and CTNNB1 improved extraovarian endometriosis lesion border detection in a relatively small cohort (n = 20), although larger studies with different endometriosis subtypes in variable cycle phases and under hormonal medication are required. The highly expressed SFRP2 and CTNNB1 improve endometriosis lesion border detection, which can have clinical implications for better visualization of endometriosis lesions over CD10. Furthermore, SFRP2 acts as a canonical WNT/CTNNB1 signaling agonist in endometriosis and positively regulates endometriosis lesion growth, suggesting that the WNT pathway may be an important therapeutic target for endometriosis. This study was funded by the Academy of Finland and by Tekes: Finnish Funding Agency for Innovation. The authors have no conflict of interest to declare.