Merck
  • Triiodothyronine modulates neuronal plasticity mechanisms to enhance functional outcome after stroke.
Triiodothyronine modulates neuronal plasticity mechanisms to enhance functional outcome after stroke.
Acta neuropathologica communications (2019-12-23)
Daniela Talhada, Joana Feiteiro, Ana Raquel Costa, Tiago Talhada, Elisa Cairrão, Tadeusz Wieloch, Elisabet Englund, Cecília Reis Santos, Isabel Gonçalves, Karsten Ruscher
ZUSAMMENFASSUNG
The development of new therapeutic approaches for stroke patients requires a detailed understanding of the mechanisms that enhance recovery of lost neurological functions. The efficacy to enhance homeostatic mechanisms during the first weeks after stroke will influence functional outcome. Thyroid hormones (TH) are essential regulators of neuronal plasticity, however, their role in recovery related mechanisms of neuronal plasticity after stroke remains unknown. This study addresses important findings of 3,5,3'-triiodo-L-thyronine (T3) in the regulation of homeostatic mechanisms that adjust excitability - inhibition ratio in the post-ischemic brain. This is valid during the first 2 weeks after experimental stroke induced by photothrombosis (PT) and in cultured neurons subjected to an in vitro model of acute cerebral ischemia. In the human post-stroke brain, we assessed the expression pattern of TH receptors (TR) protein levels, important for mediating T3 actions.Our results show that T3 modulates several plasticity mechanisms that may operate on different temporal and spatial scales as compensatory mechanisms to assure appropriate synaptic neurotransmission. We have shown in vivo that long-term administration of T3 after PT significantly (1) enhances lost sensorimotor function; (2) increases levels of synaptotagmin 1&2 and levels of the post-synaptic GluR2 subunit in AMPA receptors in the peri-infarct area; (3) increases dendritic spine density in the peri-infarct and contralateral region and (4) decreases tonic GABAergic signaling in the peri-infarct area by a reduced number of parvalbumin+ / c-fos+ neurons and glutamic acid decarboxylase 65/67 levels. In addition, we have shown that T3 modulates in vitro neuron membrane properties with the balance of inward glutamate ligand-gated channels currents and decreases synaptotagmin levels in conditions of deprived oxygen and glucose. Interestingly, we found increased levels of TRβ1 in the infarct core of post-mortem human stroke patients, which mediate T3 actions. Summarizing, our data identify T3 as a potential key therapeutic agent to enhance recovery of lost neurological functions after ischemic stroke.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly-L-Lysin -Lösung, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
SAFC
L-Glutamin
Sigma-Aldrich
L-Glutaminsäure Mononatriumsalz Monohydrat, ≥98.0% (NT)
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid