Merck
Multifunctional gap-enhanced Raman tags for preoperative and intraoperative cancer imaging.
Acta biomaterialia (2020-01-14)
Bowen Shi, Benyan Zhang, Yuqing Zhang, Yuqing Gu, Chao Zheng, Jing Yan, Weibo Chen, Fuhua Yan, Jian Ye, Huan Zhang
ZUSAMMENFASSUNG
Multi-modality imaging agents are desirable for tumor diagnosis because they can provide more alternative and reliable information for accurate detection and therapy of diseases than single imaging technique. However, most reported conventional imaging agents have not been found to successfully overcome the disadvantages of traditional diagnoses such as sensitivity, spatial resolution, short half-decay time and complexity. Therefore, exploring a multifunctional nanocomposite with the combination of their individual modality characteristics has great impact on preoperative imaging and intraoperative diagnosis of cancer. In our study, mesoporous silica gadolinium-loaded gap-enhanced Raman tags (Gd-GERTs) specifically for preoperative and intraoperative imaging are designed and their imaging capability and biosafety are examined. They exhibit strong attenuation property for computed X-ray tomography (CT) imaging, high T1 relaxivity for magnetic resonance (MR) imaging capability and surface-enhanced Raman spectroscopy (SERS) signal with good dispersity and stability, which presents CT/MR/SERS multi-mode imaging performance of the tumor of mice within a given time. Furthermore, in vivo biodistribution and long-term toxicity studies reveal that the Gd-GERTs have good biocompatibility and bio-safety. Therefore, Gd-GERTs are of great potential as a multifunctional nanoplatform for accurate preoperative CT/MRI diagnosis and intraoperative Raman imaging-guide resection of cancers. STATEMENT OF SIGNIFICANCE: Recent advances in molecular imaging technology have provided a myriad of opportunities to prepare various nanomaterials for accurate diagnosis and response evaluation of cancer via different imaging modalities. However, single bioimaging modality is still challenging to overcome the issues such as sensitivity, spatial resolution, imaging speed and complexity for clinicians. In this work, we designed a kind of unique multifunctional nanoprobes with computed X-ray tomography/magnetic resonance/surface-enhanced Raman spectroscopy (CT/MR/SERS) triple-modal imaging capabilities. Multifunctional nanotags offer the capabilities of preoperative noninvasive CT/MR imaging for identification of tumors as well as intraoperative real-time SERS imaging for guidance of complete resection of tumors. These multifunctional nanoprobes show critical clinical significance on the improvement of tumor diagnosis and therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Nitrothiophenol, technical grade, 80%