Merck
Assessing washing methods for reduction of pesticide residues in Capia pepper with LC-MS/MS.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2019-09-17)
Burak Polat, Osman Tiryaki
ZUSAMMENFASSUNG
The effects of washing treatments on removal rates of some pesticides residues (acetamiprid, chlorpyrifos and formetanate hydrochloride) on pepper were investigated. Method verification was conducted through spiking pepper samples at 0.1, 1.0 and 10.0 × MRL. QuEChERS method produced average recovery of 104.91% with relative standard deviation (RSD) of 13.41%. LOQ values of acetamiprid, chlorpyrifos and formetanate hydrochloride were estimated as 2, 10 and 5 µg/kg, respectively. Capia peppers grown in open fields were sprayed three times with pesticides. Peppers were harvested after 1st, 2nd and 3rd day of the treatments. Then the peppers were subjected to tap water, acetic acid and citric acid washing and ultrasonic cleaning treatments (for 2 and 5 min). Based on three different harvest times and two different washing durations, processing factors (PFs) and reduction rates were calculated for each washing treatment. The residues gradually decreased during washing treatments with increasing process duration. Similarly, a gradual reduction was noted with the progress of harvest times. This in turn corresponded to an increase in PF. Ultrasonic cleaning and citric acid (9%) washing were more effective than the others. Non-systemic pesticides (chlorpyrifos) were more readily removed than the systemic ones (acetamiprid). Similarly, highly soluble pesticides exhibited higher reduction.