Merck
The expression pattern of nuclear receptors during cerebellar development.
Developmental dynamics : an official publication of the American Association of Anatomists (2007-01-06)
Jun Qin, Jae Mi Suh, Bum-Jun Kim, Cheng-Tai Yu, Toshiya Tanaka, Tatsuhiko Kodama, Ming-Jer Tsai, Sophia Y Tsai
ZUSAMMENFASSUNG
The cerebellum is essential for fine control of movement and posture, and it has been a useful model for studying many aspects of neural development because of its relatively simple anatomy and developmental program. However, the roles of nuclear receptors (NRs) underlying formation of the cerebellum and maintenance of cerebellar functions are still poorly characterized. As a contribution to the Nuclear Receptor Signaling Atlas (NURSA), we employed immunohistochemistry to investigate the expression pattern of 18 NRs in the cerebellum. Ten receptors were demonstrated to be expressed in the postnatal day 21 (P21) cerebellum. Among them, five receptors (COUP-TFI, COUP-TFII, RORalpha, ERbeta, and ERRgamma) were expressed at all stages (embryonic stage, P0, P7, and P21) examined. Interestingly, COUP-TFI and COUP-TFII show differential anterior-posterior expression patterns during cerebellar development. Taken together, our results suggest that members of the nuclear receptor superfamily might play importantly physiological roles in the cerebellum.