Merck
  • Ginsenoside Rg3 suppresses the NLRP3 inflammasome activation through inhibition of its assembly.
Ginsenoside Rg3 suppresses the NLRP3 inflammasome activation through inhibition of its assembly.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-10)
Yuhua Shi, Huanan Wang, Mengjie Zheng, Wei Xu, Yang Yang, Fushan Shi
ZUSAMMENFASSUNG
Ginsenoside Rg3 is one of the main constituents of Panax ginseng. Compelling evidence has demonstrated that ginsenoside Rg3 is capable of inhibiting inflammation. However, the mechanism mediating its anti-inflammatory effects remain unclear. Here we show that ginsenoside Rg3 blocks IL-1β secretion and caspase-1 activation through inhibiting LPS priming and the NLRP3 inflammasome activation in human and mouse macrophages. Rg3 specifically inhibits activation of NLRP3 but not the NLRC4 or AIM2 inflammasomes. In addition, Rg3 has no effect on upstream regulation of NLRP3 inflammasome, such as K+ efflux, ROS production, or mitochondrial membrane potential. Mechanistically, Rg3 abrogates NEK7-NLRP3 interaction, and subsequently inhibits NLRP3-ASC interaction, ASC oligomerization, and speckle formation. More importantly, Rg3 can reduce IL-1β secretion induced by LPS in mice and protect mice from lethal endotoxic shock. Thus, our findings reveal an anti-inflammatory mechanism for Rg3 and suggest its potential use in NLRP3-driven diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Ginsenosid Rg3, ≥98% (HPLC)
Sigma-Aldrich
Bernsteinsäure bis(N-Hydroxysuccinimid-Ester), ≥95%, powder
Millipore
ProteinA-Sepharose 4, Fast Flow aus Staphylococcus aureus, aqueous ethanol suspension
Millipore
FLAG® M Purification Kit, For Mammalian expression systems.