Merck
  • Kisspeptin-GPR54 signaling is essential for preovulatory gonadotropin-releasing hormone neuron activation and the luteinizing hormone surge.
Kisspeptin-GPR54 signaling is essential for preovulatory gonadotropin-releasing hormone neuron activation and the luteinizing hormone surge.
The Journal of Neuroscience (2008)
Clarkson J, d'Anglemont de Tassigny X, et al.