Merck
  • Local application of zoledronic acid incorporated in a poly (D, L-lactide)-coated implant accelerates fracture healing in rats
Local application of zoledronic acid incorporated in a poly (D, L-lactide)-coated implant accelerates fracture healing in rats
Acta Orthopaedica (2008)
Greiner, S.H. et al.