Merck
Blockade of N-Glycosylation Promotes Antitumor Immune Response of T Cells.
Journal of immunology (Baltimore, Md. : 1950) (2020-01-24)
Shigemi Sasawatari, Yuki Okamoto, Atsushi Kumanogoh, Toshihiko Toyofuku
ZUSAMMENFASSUNG
Adoptive cellular therapy and its derivative, chimeric AgR T cell therapy, have achieved significant progress against cancer. Major barriers persist, however, including insufficient induction of cytotoxic T cells and exhaustion of tumor-infiltrating lymphocytes. In this study, we discovered a new role for 2-deoxy-d-glucose (2DG) in enhancing the antitumor activity of human T cells against NKG2D ligand-expressing tumor cells. Human T cells treated with 2DG upregulated the NK-specific transcription factors TOX2 and EOMES, thereby acquiring NK cell properties, including high levels of perforin/granzyme and increased sensitivity to IL-2. Notably, rather than inhibiting glycolysis, 2DG modified N-glycosylation, which augmented antitumor activity and cell surface retention of IL-2R of T cells. Moreover, 2DG treatment prevented T cells from binding to galectin-3, a potent tumor Ag associated with T cell anergy. Our results, therefore, suggest that modifying N-glycosylation of T cells with 2DG could improve the efficacy of T cell-based immunotherapies against cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Laktose, ≤30% α-anomer basis, ≥99% total lactose basis
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
Natrium-Oxamat, ≥98%
Sigma-Aldrich
Brombrenztraubensäure, ≥98.0%
Sigma-Aldrich
(2S,3S,4R)-1-O-(α-D-Galactosyl)-N-hexacosanoyl-2-amino-1,3,4-octadecantriol, ≥95% (TLC)
Sigma-Aldrich
Bovine IFNG / Interferon Gamma ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse Granzyme B ELISA Kit, for serum, plasma and cell culture supernatant