Merck
  • Proline derivatives in fruits of bergamot (Citrus bergamia Risso et Poit): presence of N-methyl-L-proline and 4-hydroxy-L-prolinebetaine.
Proline derivatives in fruits of bergamot (Citrus bergamia Risso et Poit): presence of N-methyl-L-proline and 4-hydroxy-L-prolinebetaine.
Journal of agricultural and food chemistry (2010-12-07)
Luigi Servillo, Alfonso Giovane, Maria Luisa Balestrieri, Domenico Cautela, Domenico Castaldo
ZUSAMMENFASSUNG
The content of proline and various compounds deriving from its metabolism (4-hydroxy-L-proline, N-methyl-L-proline, N,N-dimethylproline, and 4-hydroxy-L-prolinebetaine) was determined in fruits and seeds of Bergamot (Citrus bergamia Risso et Poit), growing in the Calabria region (South Italy). A HPLC-ESI-tandem mass spectrometry method, which allowed rapid determination of L-proline, 4-hydroxy-L-proline, N-methyl-L-proline, N,N-dimethylproline, and 4-hydroxy-L-prolinebetaine in juice and extracts of bergamot fruit with minimum sample preparation and short analysis time (about 10 min), is presented. Proline and 4-hydroxy-L-proline levels in the samples were also determined by HPLC analysis with fluorescence detection and the results compared to those obtained with HPLC-ESI-tandem mass spectrometry. For the first time, the presence of N-methyl-L-proline and 4-hydroxy-L-prolinebetaine in the fruits of a plant of the Citrus genus is reported.