Merck
  • New insights into the coordination of Cu(II) by the amyloid-B 16 peptide from Fourier transform IR spectroscopy and isotopic labeling.
New insights into the coordination of Cu(II) by the amyloid-B 16 peptide from Fourier transform IR spectroscopy and isotopic labeling.
The journal of physical chemistry. B (2011-10-27)
Youssef El Khoury, Pierre Dorlet, Peter Faller, Petra Hellwig
ZUSAMMENFASSUNG
Alzheimer's disease is a neurodegenerative disorder in which the formation of amyloid-β (Aβ) aggregates plays a causative role. There is ample evidence that Cu(II) can bind to Aβ and modulate its aggregation. Moreover, Cu(II) bound to Aβ might be involved in the production of reactive oxygen species, a process supposed to be involved in the Alzheimer's disease. The native Aβ40 contains a high affinity binding site for Cu(II), which is comprised in the N-terminal portion. Thus, Aβ16 (amino acid 1-16 of Aβ) has often been used as a model for Cu(II)-binding to monomeric Aβ. The Cu(II)-binding to Aβ is pH dependent and at pH 7.4, two different type of Cu(II) coordinations exist in equilibrium. These two forms are predominant at pH 6.5 and pH 9.0. In either form, a variety of studies show that the N-terminal Asp and the three His play a key role in the coordination, although the exact binding of these amino acids has not been addressed. Therefore, we studied the coordination modes of Cu(II) at pH 6.5 and 9.0 with the help of Fourier transform infrared (FTIR) spectroscopy. Combined with isotopic labeling of the amino acids involved in the coordination sphere, the data points toward the coordination of Cu(II) via the carboxylate of Asp1 at both pH values in a pseudobridging monovalent fashion. At low pH, His6 binds copper via Nτ, while His13 and His14 are bound via Nπ. At high pH, direct evidence is given on the coordination of Cu(II) via the Nτ atom of His6. Additionally, this study clearly shows the effect of Cu(II) binding on the protonation state of the His residues where a proton displacement takes places on the nitrogen atoms of the imidazole ring.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
[Glu11]-Amyloid β 1-16