Merck
  • Endothelin-converting enzyme-1 actions determine differential trafficking and signaling of corticotropin-releasing factor receptor 1 at high agonist concentrations.
Endothelin-converting enzyme-1 actions determine differential trafficking and signaling of corticotropin-releasing factor receptor 1 at high agonist concentrations.
Molecular endocrinology (Baltimore, Md.) (2012-02-11)
Burcu Hasdemir, Shilpi Mahajan, Nigel W Bunnett, Min Liao, Aditi Bhargava
ZUSAMMENFASSUNG
CRF receptor 1 (CRF(1)), a key neuroendocrine mediator of the stress response, has two known agonists corticotropin-releasing factor (CRF) and urocortin 1 (Ucn1). Here we report that endothelin-converting enzyme-1 (ECE-1) differentially degrades CRF and Ucn1; ECE-1 cleaves Ucn1, but not CRF, at critical residue Arginine-34/35', which is essential for ligand-receptor binding. At near K(D) agonist concentration (30 nm), both Ucn1- and CRF-mediated Ca(2+) mobilization are ECE-1 dependent. Interestingly, at high agonist concentration (100 nm), Ucn1-mediated Ca(2+) mobilization remains ECE-1 dependent, whereas CRF-mediated mobilization becomes independent of ECE-1 activity. At high agonist concentration, ECE-1 inhibition disrupted Ucn1-, but not CRF-induced CRF(1) recycling and resensitization, but did not prolong the association of CRF(1) with β-arrestins. RNA interference-mediated knockdown of Rab suggests that both Ucn1- and CRF-induced CRF(1) resensitization is dependent on activity of Rab11, but not of Rab4. CRF(1) behaves like a class A G protein-coupled receptor with respect to transient β-arrestins interaction. We propose that differential degradation by ECE-1 is a novel mechanism by which CRF(1) receptor is protected from overactivation by physiologically relevant high concentrations of higher affinity ligand to mediate distinct resensitization and downstream signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
ECE-1 Substrate, ≥95% (HPLC)