Merck
  • Excitatory pathway engaging glutamate, calcineurin, and NFAT upregulates IL-4 in ischemic neurons to polarize microglia.
Excitatory pathway engaging glutamate, calcineurin, and NFAT upregulates IL-4 in ischemic neurons to polarize microglia.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2019-03-21)
Shun-Ming Ting, Xiurong Zhao, Xueping Zheng, Jaroslaw Aronowski
ZUSAMMENFASSUNG
Excitotoxicity and microglia/macrophage over-activation are the important pathogenic steps in brain damage caused by ischemic stroke. Recent studies from our group suggest that the neurons in ischemic penumbra generate an anti-inflammatory cytokine, interleukin-4 (IL-4). This neuron-produced IL-4 could subsequently convert surrounding microglia/macrophages to a reparative (M2)-phenotype. The present study was designed to establish the mechanisms by which neurons under transient ischemic condition produce/secrete IL-4. We employed primary rat cortical neurons and a validated in vitro ischemic injury model involving transient oxygen-glucose deprivation (OGD). We discovered that only sublethal OGD induces IL-4 production/secretion by neurons. We then showed that excitotoxic stimulus (an integral component of OGD-mediated damage) involving N-methyl-D-aspartate (NMDA), and not kainate receptor, triggers neuronal IL-4 production/release. Of note, oxidative stress or pro-apoptotic stimuli did not induce IL-4 production by neurons. Next, using the calcineurin inhibitor FK506, we implicated this phosphatase in activation of the nuclear factor of activated T-cells (NFAT; a transcription factor activated through calcineurin-mediated dephosphorylation) and propose that this pathway is involved in transcriptional upregulation of the IL-4 synthesis in NMDA-treated neurons. Finally, using a transfer of culture medium from NMDA-conditioned neuron to microglia, we showed that the neuronal IL-4 can polarize microglia toward a restorative, phagocytic phenotype.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid