- Discovery of the first M5-selective and CNS penetrant negative allosteric modulator (NAM) of a muscarinic acetylcholine receptor: (S)-9b-(4-chlorophenyl)-1-(3,4-difluorobenzoyl)-2,3-dihydro-1H-imidazo[2,1-a]isoindol-5(9bH)-one (ML375).
Discovery of the first M5-selective and CNS penetrant negative allosteric modulator (NAM) of a muscarinic acetylcholine receptor: (S)-9b-(4-chlorophenyl)-1-(3,4-difluorobenzoyl)-2,3-dihydro-1H-imidazo[2,1-a]isoindol-5(9bH)-one (ML375).
Journal of medicinal chemistry (2013-10-30)
Patrick R Gentry, Masaya Kokubo, Thomas M Bridges, Nathan R Kett, Joel M Harp, Hyekyung P Cho, Emery Smith, Peter Chase, Peter S Hodder, Colleen M Niswender, J Scott Daniels, P Jeffrey Conn, Michael R Wood, Craig W Lindsley
PMID24164599
ZUSAMMENFASSUNG
A functional high throughput screen and subsequent multidimensional, iterative parallel synthesis effort identified the first muscarinic acetylcholine receptor (mAChR) negative allosteric modulator (NAM) selective for the M5 subtype. ML375 is a highly selective M5 NAM with submicromolar potency (human M5 IC50 = 300 nM, rat M5 IC50 = 790 nM, M1-M4 IC50 > 30 μM), excellent multispecies PK, high CNS penetration, and enantiospecific inhibition.
MATERIALIEN