Merck
  • Reactivity of charged LiVPO4F with 1 M LiPF6 EC: DEC electrolyte at high temperature as studied by accelerating rate calorimetry
Reactivity of charged LiVPO4F with 1 M LiPF6 EC: DEC electrolyte at high temperature as studied by accelerating rate calorimetry
Electrochemical Communications (2009)
Zhou F, et al.