Merck
Abl-mediated PI3K activation regulates macrophage podosome formation.
Journal of cell science (2020-05-13)
Yuhuan Zhou, Zhen Feng, Fakun Cao, Xiaoting Liu, Xiaojie Xia, Cheng-Han Yu
ZUSAMMENFASSUNG
Podosomes play crucial roles in macrophage adhesion and migration. Wiskott-Aldrich syndrome protein (WASP; also known as WAS)-mediated actin polymerization is one of the key events initiating podosome formation. Nevertheless, membrane signals to trigger WASP activation at macrophage podosomes remain unclear. Here, we show that phosphatidylinositol (3,4,5)-trisphosphate [PI(3,4,5)P3] lipids are enriched at the podosome and stably recruit WASP rather than the WASP-5KE mutant. Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit β (PIK3CB) is spatially located at the podosome core. Inhibition of PIK3CB and overexpression of phosphatase and tensin homolog (PTEN) impede F-actin polymerization of the podosome. PIK3CB activation is regulated by Abl1 and Src family kinases. At the podosome core, Src and Hck promote the phosphorylation of Tyr488 in the consensus Y-x-x-M motif of Abl1, which enables the association of phosphoinositide 3-kinase (PI3K) regulatory subunits. Knockdown of Abl1 rather than Abl2 suppresses the PI3K/Akt pathway, regardless of Src and Hck activities. Reintroduction of wild-type Abl1 rather than the Abl1-Y488F mutant rescues PI3KR1 recruitment and PI3K activation. When PIK3CB, Abl1 or Src/Hck is suppressed, macrophage podosome formation, matrix degradation and chemotactic migration are inhibited. Thus, Src/Hck-mediated phosphorylation of Abl1 Tyr488 triggers PIK3CB-dependent PI(3,4,5)P3 production and orchestrates the assembly and function of macrophage podosomes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polybren Infektions- / Transfektionsreagenz, A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.
Sigma-Aldrich
Anti-Vinculin-Antikörper, Maus monoklonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
Imatinib
Sigma-Aldrich
PP2, ≥98% (HPLC)