Merck
  • Antitumor Activity of an Immobilized Peroxidase System Against Murine Ehrlich Ascites Is Mediated by the Immune System
Antitumor Activity of an Immobilized Peroxidase System Against Murine Ehrlich Ascites Is Mediated by the Immune System
Exp. Biol. Med (1993)
Steven H. Combs, Clinton D. Morgan, et al.