Merck
Digestion of carbohydrate from white beans (Phaseolus vulgaris L.) in healthy humans.
The Journal of nutrition (1998-06-18)
L Noah, F Guillon, B Bouchet, A Buléon, C Molis, M Gratas, M Champ
ZUSAMMENFASSUNG
Resistant starch (RS) is thought to be present in large amounts in legume seeds; however, it has never been quantified in healthy humans. RS from cooked (atmospheric pressure) white beans was quantified in humans and pigs, and characterized to explain its low digestibility. Six human volunteers were intubated to collect ileal digesta after an experimental meal composed of orange juice, butter and 167 g beans (dry matter basis). The reliability of the intubation method was examined in a pig study in which it was compared with another collection method, ileal cannulation. Chemical analyses, microscopy and size exclusion chromatography were performed on human and pig digesta. The pig study showed that the intubation method may underestimate the quantity of RS. However, no chemical/physical difference was observed between the RS collected by the two techniques. In the human study, 16.5 +/- 1.3% (11.3 g) of the ingested starch was recovered as RS. The microscopy of the digesta showed that part of the RS was enclosed in the cell walls. Although the RS was composed mainly of alpha-glucan molecules with a degree of polymerization (DP) 40 to 60, oligosaccharides and large molecules of DP > 400 were also present. Retrogradation was not found to be the main factor responsible for starch malabsorption. We conclude that white beans may contain a large amount of RS formed mainly by partially degraded molecules protected by the cell walls during their transit through the gut.