Merck
  • Benign preparation of metal-organic frameworks of trimesic acid and Cu, Co or Ni for potential sensor applications
Benign preparation of metal-organic frameworks of trimesic acid and Cu, Co or Ni for potential sensor applications
Journal of Electronic Materials (2015)
Sel K, et al.