- XY aneuploidy rate in sperm of two ?minor? breeds of cattle (Bos taurus) by using dual color fluorescent in situ hybridization (FISH)
XY aneuploidy rate in sperm of two ?minor? breeds of cattle (Bos taurus) by using dual color fluorescent in situ hybridization (FISH)
Theriogenology (2010)
Pauciullo, Alfredo, et al