Merck
  • Podosome formation promotes plasma membrane invagination and integrin-β3 endocytosis on a viscous RGD-membrane.
Podosome formation promotes plasma membrane invagination and integrin-β3 endocytosis on a viscous RGD-membrane.
Communications biology (2020-03-15)
Fakun Cao, Yuhuan Zhou, Xiaoting Liu, Cheng-Han Yu
ZUSAMMENFASSUNG
Integrin receptors orchestrate cell adhesion and cytoskeletal reorganization. The endocytic mechanism of integrin-β3 receptor at the podosome remains unclear. Using viscous RGD-membrane as the model system, here we show that the formation of podosome-like adhesion promotes Dab2/clathrin-mediated endocytosis of integrin-β3. Integrin-β3 and RGD ligand are endocytosed from the podosome and sorted into the endosomal compartment. Inhibitions of podosome formation and knockdowns of Dab2 and clathrin reduce RGD endocytosis. F-actin assembly at the podosome core exhibits protrusive contact towards the substrate and results in plasma membrane invaginations at the podosome ring. BIN1 specifically associates with the region of invaginated membrane and recruits DNM2. During the podosome formation, BIN1 and DNM2 synchronously enrich at the podosome ring and trigger clathrin dissociation and RGD endocytosis. Knockdowns of BIN1 and DNM2 suppress RGD endocytosis. Thus, plasma membrane invagination caused by F-actin polymerization promotes BIN1-dependent DNM2 recruitment and facilitate integrin-β3 endocytosis at the podosome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dulbecco Modifiziertes Eagle-Medium – hoher Glucosegehalt, HEPES Modification, With 4500 mg/L glucose, L-glutamine, and 25 mM HEPES, without sodium bicarbonate and pyruvate, powder, suitable for cell culture
Sigma-Aldrich
1,2-Dioleoyl-sn-Glycero-3-Phosphocholin, lyophilized powder
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
Anti-LIBS2 epitope (Integrin beta3 subunit) Antibody, clone ab62, clone ab62, from mouse
Sigma-Aldrich
Anti-Bin1 Antibody, clone 99D, clone 99D, Upstate®, from mouse