Merck
  • Profiling of Flavonoid and Antioxidant Activity of Fruit Tissues from 27 Chinese Local Citrus Cultivars.
Profiling of Flavonoid and Antioxidant Activity of Fruit Tissues from 27 Chinese Local Citrus Cultivars.
Plants (Basel, Switzerland) (2020-02-09)
Qiyang Chen, Dan Wang, Chun Tan, Yan Hu, Balasubramani Sundararajan, Zhiqin Zhou
ZUSAMMENFASSUNG
Flavonoid profile and antioxidant activity of citrus peels, pulps, and juices from 27 local citrus cultivars in China were investigated. Flavonoid composition and content were determined using UPLC-PDA. Total phenolic content (TPC) and total flavonoid content (TFC) were measured using a Folin-Ciocalteau reagent and Al(NO3)-NaNO2 complexometry, respectively. The antioxidant capacities of the extracts were evaluated by DPPH, ABTS and FRAP method, respectively. Citrus peel not only exhibited better antioxidant potential, but also presented more composition diversity and contained higher concentrations of flavonoids than pulp and juice. Different citrus species were characterized by their individual predominant flavonoids, contributing largely to the antioxidant activity, such as mandarin was characterized by hesperidin, nobiletin and tangeretin, while pummelo and papeda were characterized by naringin. The peel of Guihuadinanfeng (Citrus reticulata) had the highest TPC of 23.46 mg equivalent gallic acid/g DW (dry weight) and TFC of 21.37 mg equivalent rutin/g DW. Shiyueju (C.reticulata) peel showed the highest antioxidant capacity based on the antioxidant potency composite (APC) analysis. Overall, mandarin (C. reticulata) fruits peel contained more TPC and TFC, exhibiting higher antioxidant capacities than other species, and were good natural sources of flavonoids and antioxidants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carbonsäure, 97%
Sigma-Aldrich
Narirutin, ≥98% (HPLC)