Merck
  • ATR signalling mediates the prosurvival function of phospho-NPM against PIDDosome mediated cell death.
ATR signalling mediates the prosurvival function of phospho-NPM against PIDDosome mediated cell death.
Cellular signalling (2020-03-21)
Disha Hiregange, Hemanth Naick, Basuthkar J Rao
ZUSAMMENFASSUNG
We uncover a novel non-canonical function of ATR kinase in the control of PIDDosome activation, and show that under normal cellular conditions involving no replication stress, ATR kinase controls the phosphorylation of cellular NPM via pChk1 as well as the two phosphatases, PPM1D and PP1β. We show that pNPM triggers the dissociation of NPM from PIDD, preventing the cell from undergoing caspase 2 mediated cell death via PIDDosome, thereby acting as an endogenous negative regulator of PIDDosome activation. pChk1 interaction with NPM is abrogated following ATR kinase inhibition, leading to the drop in nucleoplasmic/chromatin pNPM level, inducing PIDD. Consistent with this mechanism, the phosphomimic mutants of Chk1 and NPM become refractory to ATR/pChk1 kinase inhibition by avoiding PIDDosome signalling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting human CHEK1