Merck
  • GASP-2 overexpressing mice exhibit a hypermuscular phenotype with contrasting molecular effects compared to GASP-1 transgenics.
GASP-2 overexpressing mice exhibit a hypermuscular phenotype with contrasting molecular effects compared to GASP-1 transgenics.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-22)
Alexis Parenté, Axel Boukredine, Fabienne Baraige, Nathalie Duprat, Victor Gondran-Tellier, Laetitia Magnol, Véronique Blanquet
ZUSAMMENFASSUNG
Muscle atrophy is associated with many diseases including genetic disorders, sarcopenia, or cachexia syndromes. Myostatin (Mstn), a transforming growth factor-beta (TGF-β) member, plays a key role in skeletal muscle homeostasis as a powerful negative regulator. Over the last decade, about 15 clinical trials aimed at inhibiting the Mstn pathway, failed to produce conclusive results. In this context, we investigated whether growth and differentiation factor-associated serum protein-1 (GASP-1) or GASP-2, two natural inhibitors of Mstn, might represent a potential therapeutic. As we previously reported, mice overexpressing Gasp-1 (Tg(Gasp-1)) present an increase of muscle mass but develop metabolic disorders with aging. Here, we showed that overexpression of Gasp-2 increases the muscular mass without metabolic defects. We also found that Tg(Gasp-2) mice displayed, like Mstn-/- mice, a switch from slow- to fast-twitch myofibers whereas Tg(Gasp-1) mice exhibit a reverse switch. Our studies supported the fact that GASP-2 has less affinity than GASP-1 for Mstn, leading to a constitutive Mstn upregulation only in Tg(Gasp-1) mice, responsible for the observed phenotypic differences. Altogether, our findings highlighted a gene expression regulatory network of TGF-β members and their inhibitors in muscle.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Protease aus Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
Anti-Laminin in Kaninchen hergestellte Antikörper, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Human GPRASP2 / G-Protein Coupled Receptor-associated Sorting Protein 2 ELISA Kit