Merck
Predicting intestinal recovery after necrotizing enterocolitis in preterm infants.
Pediatric research (2019-10-28)
Sara J Kuik, Willemien S Kalteren, Mirthe J Mebius, Arend F Bos, Jan B F Hulscher, Elisabeth M W Kooi
ZUSAMMENFASSUNG
Intestinal recovery after NEC is difficult to predict in individuals. We evaluated whether several biomarkers predict intestinal recovery after NEC in preterm infants. We measured intestinal tissue oxygen saturation (rintSO2) and collected urinary intestinal-fatty acid binding protein (I-FABPu) levels 0-24 h and 24-48 h after NEC onset, and before and after the first re-feed. We assessed intestinal recovery in two ways: time to full enteral feeding (FEFt; below or equal/above group's median) and development of post-NEC complications (recurrent NEC/post-NEC stricture). We determined whether the rintSO2, its range, and I-FABPu differed between groups. We included 27 preterm infants who survived NEC (Bell's stage ≥ 2). Median FEFt was 14 [IQR: 12-23] days. Biomarkers only predicted intestinal recovery after the first re-feed. Mean rintSO2 ≥ 53% combined with mean rintSO2range ≥ 50% predicted FEFt < 14 days with OR 16.7 (CI: 2.3-122.2). The rintSO2range was smaller (33% vs. 51%, p < 0.01) and I-FABPu was higher (92.4 vs. 25.5 ng/mL, p = 0.03) in case of post-NEC stricture, but not different in case of recurrent NEC, compared with infants without complications. The rintSO2, its range, and I-FABPu after the first re-feed after NEC predicted intestinal recovery. These biomarkers have potential value in individualizing feeding regimens after NEC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Human FABP2 / Fatty Acid-Binding Protein, Intestinal ELISA Kit, for serum, plasma, cell culture supernatants and urine