Merck
  • High-mobility group box 1 protein (HMGB1) from Cherry Valley duck mediates signaling pathways and antiviral activity.
High-mobility group box 1 protein (HMGB1) from Cherry Valley duck mediates signaling pathways and antiviral activity.
Veterinary research (2020-02-20)
Xiaolan Hou, Gen Liu, Huihui Zhang, Xiaofang Hu, Xinyue Zhang, Fei Han, Huizhen Cui, Jinjian Luo, Ru Guo, Rong Li, Ning Li, Liangmeng Wei
ZUSAMMENFASSUNG
High-mobility group box 1 protein (HMGB1) shows endogenous damage-associated molecular patterns (DAMPs) and is also an early warning protein that activates the body's innate immune system. Here, the full-length coding sequence of HMGB1 was cloned from the spleen of Cherry Valley duck and analyzed. We find that duck HMGB1(duHMGB1) is mostly located in the nucleus of duck embryo fibroblast (DEF) cells under normal conditions but released into the cytoplasm after lipopolysaccharide (LPS) stimulation. Knocking-down or overexpressing duHMGB1 had no effect on the baseline apoptosis rate of DEF cells. However, overexpression increased weakly apoptosis after LPS activation. In addition, overexpression strongly activated the IFN-I/IRF7 signaling pathway in DEF cells and significantly increased the transcriptional level of numerous pattern recognition receptors (PRRs), pro-inflammatory cytokines (IL-6, TNF-α), IFNs and antiviral molecules (OAS, PKR, Mx) starting from 48 h post-transfection. Overexpression of duHMGB1 strongly impacted duck virus replication, either by inhibiting it from the first stage of infection for novel duck reovirus (NDRV) and at late stage for duck Tembusu virus (DTMUV) or duck plague virus (DPV), or promoting replication at early stage for DTMUV and DPV infection. Importantly, data from duHMGB1 overexpression and knockdown experiments, time-dependent DEF cells transcriptional immune responses suggest that duHMGB1 and RIG-I receptor might cooperate to promote the expression of antiviral proteins after NDRV infection, as a potential mechanism of duHMGB1-mediated antiviral activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Octoclothepin maleate salt, solid