Merck
  • Preparation and Characterization of Epoxy Resin Filled with Ti3C2Tx MXene Nanosheets with Excellent Electric Conductivity.
Preparation and Characterization of Epoxy Resin Filled with Ti3C2Tx MXene Nanosheets with Excellent Electric Conductivity.
Nanomaterials (Basel, Switzerland) (2020-01-23)
Ailing Feng, Tianqi Hou, Zirui Jia, Yi Zhang, Fan Zhang, Guanglei Wu
ZUSAMMENFASSUNG
MXene represents new kinds of two-dimensional material transition metal carbides and/or carbonitrides, which have attracted much attention in various applications including electrochemical storage devices, catalysts, and polymer composite. Here, we report a facile method to synthesize Ti3C2Tx MXene nanosheets and prepare a novel electrically conductive adhesive based on epoxy resin filled with Ti3C2Tx MXene nanosheets by solution blending. The structure, morphology, and performance of Ti3C2Tx MXene nanosheets and epoxy/Ti3C2Tx MXene nanosheets composite were investigated. The results show that Ti3C2Tx MXene possesses nanosheet structure. Ti3C2Tx MXene nanosheets were homogeneously dispersed in epoxy resin. Electrical conductivity and mechanical properties measurements reveal that the epoxy/Ti3C2Tx MXene nanosheet composite exhibited both good electrical conductivity (4.52 × 10-4 S/m) and favorable mechanical properties (tensile strength of 66.2 MPa and impact strength of 24.2 kJ/m2) when the content of Ti3C2Tx MXene nanosheets is 1.2 wt %. Thus, Ti3C2Tx MXene is a promising filler for electrically conductive adhesive with high electric conductivity and high mechanical performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D.E.R. 332, used as embedding medium