Merck
  • SET domain-containing 5 is a potential prognostic biomarker that promotes esophageal squamous cell carcinoma stemness.
SET domain-containing 5 is a potential prognostic biomarker that promotes esophageal squamous cell carcinoma stemness.
Experimental cell research (2020-01-26)
Lihua Piao, Haoyue Li, Ying Feng, Zhaoting Yang, Seokhyung Kim, Yanhua Xuan
ZUSAMMENFASSUNG
SET domain-containing 5 (SETD5) is an uncharacterized member of the protein lysine methyltransferase family. Although it was reported that SETD5 gene mutations are associated with the several types of human cancer, its functional role in esophageal squamous cell carcinoma (ESCC) progression has not been fully elucidated. In the present study, we used tissue samples from 147 patients with ESCC and ESCC cell lines to determine the clinicopathological significance of SETD5 in ESCC and its effects on ESCC stemness. We performed immunohistochemical staining, immunofluorescence imaging, and tumor sphere formation, colony formation, flow cytometry, wound healing, Transwell, and western blotting assays. SETD5 expression was upregulated in ESCC tissue and associated with primary tumor (pT) stage, clinical stage, lymph node metastasis, shorter overall survival rate, and disease-free survival rate. Cox regression analyses indicated that SETD5 is an independent poor prognostic factor of ESCC. In addition, SETD5 expression was correlated with cancer stemness-related protein, hypoxia-inducible factor-1α (HIF-1α), and CD68 expression. Moreover, immunofluorescence analysis revealed that SETD5 was co-localized with CD44 and SOX2 in TE10 and TE11 cells and that exposing cells to cobalt chloride increased HIF-1α, SETD5, and stemness-related protein expression in a time-dependent manner. Furthermore, SETD5 expression was significantly correlated with the expression of cell cycle-related genes and PI3K/Akt signaling pathway-related proteins. Finally, knocking down SETD5 downregulated the expression of stemness-related and PI3K/Akt signaling pathway proteins, while inhibiting tumor spheroid formation, cell proliferation, migration, and invasion in ESCC cells. These results indicate that SETD5 expression is associated with cancer stemness and that SETD5 is a potential prognostic biomarker and therapeutic target for ESCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human SETD5