Merck
  • Upregulation of microRNA-204 improves insulin resistance of polycystic ovarian syndrome via inhibition of HMGB1 and the inactivation of the TLR4/NF-κB pathway.
Upregulation of microRNA-204 improves insulin resistance of polycystic ovarian syndrome via inhibition of HMGB1 and the inactivation of the TLR4/NF-κB pathway.
Cell cycle (Georgetown, Tex.) (2020-02-25)
Bin Jiang, Min Xue, Dabao Xu, Yujia Song, Shujuan Zhu
ZUSAMMENFASSUNG
There is growing evidence of the position of microRNAs (miRs) in polycystic ovarian syndrome (PCOS), thus our objective was to discuss the impact of miR-204 on insulin resistance (IR) in PCOS by targeting highmobility group box protein 1(HMGB1)-mediated toll-like receptor 4(TLR4)/nuclear factor-kappa B (NF-κB) pathway.PCOS-IR patients and PCOS non-insulin resistance (PCOS-NIR) patients were included. The levels of serum sex hormones and related insulin were measured, the expression of miR-204, HMGB1, TLR4 and NF-κB p65 was detected, the diagnostic efficacy of miR-204 in PCOS-IR was analyzed, and the correlation between the expression of miR-204 in PCOS-IR and fasting blood glucose (FPG), fasting insulin (FINS), homeostasis model of assessment for insulin resistance index (HOMA-IR) was analyzed. Both in vitro and in vivo experiments were performed to elucidate the capabilities of miR-204 and HMGB1 in proliferation and apoptosis of PCOS-IR granulosa cells.MiR-204 was lowly expressed as well as HMGB1, TLR4 and NF-κB p65 were highly expressed in PCOS-IR patients. Follicule-stimulating hormone was downregulated, while luteinizing hormone, estrogen, progesterone, FPG, FINS and HOMA-IR were elevated in PCOS-IR. Upregulation of miR-204 and downregulation of HMGB1 could repress TLR4/NF-κB pathway activation, degraded insulin release and testosterone (T) leveland ascended ovarian coefficient, boosted cell proliferation and restrained apoptosis of granulosa cells. Overexpression of HMGB1 reverses the effect of upregulation of miR-204 on IR of PCOS.Our study presents that high expression of miR-204 or inhibition of HMGB1 can improve IR of PCOS via the inactivation of TLR4/NF-κB pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1