Merck
Efficient and Stable CdSe/CdS/ZnS Quantum Rods-in-Matrix Assembly for White LED Application.
Nanomaterials (Basel, Switzerland) (2020-02-16)
Yujuan Chen, Weishuo Xing, Yixuan Liu, Xinsu Zhang, Yangyang Xie, Chongyu Shen, Jay Guoxu Liu, Chong Geng, Shu Xu
ZUSAMMENFASSUNG
CdSe/CdS core-shell quantum rods (QRs) are a promising prospect in optoelectronic applications but usually have a relatively low quantum efficiency and stability. Here, we report on an efficient and stable CdSe/CdS/ZnS QRs-in-matrix assembly (QRAs) by growing and embedding CdSe/CdS QRs in ZnS matrices. Structural characterizations show that the CdSe/CdS QRs are encapsulated and interconnected by ZnS in the QRAs structure. The stable ZnS encapsulation renders the CdSe/CdS QRs high quantum efficiency (QE) up to 85%. The QRAs also present high photo- and thermal-stability and can preserve 93% of the initial QE at 100 °C. The QRAs powder presents a light degradation of only 2% under continuous excitation for 100 h, displaying profound potential in optoelectronic applications. White light-emitting diodes (WLEDs) are fabricated by packaging the QRAs powder as phosphor on top of blue GaN chip. The WLED shows high optical performance and light quality.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Octadecen, technical grade, 90%