Merck
  • Atorvastatin inhibits renal inflammatory response induced by calcium oxalate crystals via inhibiting the activation of TLR4/NF-κB and NLRP3 inflammasome.
Atorvastatin inhibits renal inflammatory response induced by calcium oxalate crystals via inhibiting the activation of TLR4/NF-κB and NLRP3 inflammasome.
IUBMB life (2020-02-23)
Yan Sun, Yunlong Liu, Xiaofeng Guan, Juening Kang, Xiang Wang, Quan Liu, Derong Li, Hua Xu, Zhiwei Tao, Yaoliang Deng
ZUSAMMENFASSUNG
This study aimed to investigate the renal protective effect of atorvastatin (ATV) on the kidney inflammation induced by calcium oxalate (CaOx) crystals. A cell model of cell-crystal interactions and a rat model of CaOx kidney stone were established. The expressions of TLR4, NF-κB, NLRP3, and cleaved caspase-1 in cells and rat kidney tissues were detected using Western blot, immunohistochemical, and/or immunofluorescence. The concentrations of malondialdehyde (MDA), superoxide dismutase (SOD), reactive oxygen species (ROS) in cells, and lactic acid dehydrogenase (LDH) in the culture medium were measured. The secreted levels of interleukin (IL)-1β, IL-18, IL-6, and tumor necrosis factor-α (TNF-α) were examined by ELISA. The serum levels of creatinine (CRE) and blood urea nitrogen (BUN) were measured. von Kossa staining was used for the evaluation of renal lens deposition. The CaOx model group showed significantly decreased SOD level; increased concentrations of MDA; ROS and LDH; elevated expressions of TLR4, NF-κB, NLRP3, and cleaved caspase-1; and the elevated release of IL-1β, IL-18, IL-6, and TNF- α as compared to the control group. The treatment with ATV significantly inhibited the formation of CaOx kidney stone by increasing the level of SOD; downregulating MDA, ROS, and LDH; inhibiting the expressions of TLR4, NF-κB, NLRP3 and cleaved caspase-1; and blocking the secretion of inflammatory cytokines. In addition, the serum levels of CRE and BUN, and the intrarenal crystal deposition were also significantly decreased in ATV-treated rats. In summary, oxidative stress, TLR4/NF-κB, and NLRP3 inflammasome pathways are involved in renal inflammatory responses induced by CaOx crystals. ATV treatment significantly suppressed oxidative stress, inhibited the activation of TLR4/NF-κB and NLRP3 inflammasome pathways, and decreased the release of inflammatory mediators, thereby ameliorating CaOx crystal-induced damage and crystal deposition in HK-2 cells and rat kidney tissues.