Merck
  • The immunomodulatory effects of mesenchymal stem cells on long term pulmonary complications in an animal model exposed to a sulfur mustard analog.
The immunomodulatory effects of mesenchymal stem cells on long term pulmonary complications in an animal model exposed to a sulfur mustard analog.
International immunopharmacology (2019-11-27)
Somaye Sadeghi, Nariman Mosaffa, Seyed Mahmoud Hashemi, Mohammad Mehdi Naghizadeh, Tooba Ghazanfari
ZUSAMMENFASSUNG
Sulfur Mustard (SM) is one of the most lethal chemicals with major complications manifested in the lungs. Although the pathogenesis behind SM-induced lung injury still remains poorly understood, prolonged activation and the imbalance of two major macrophage populations (M1 and M2) have been suggested to be involved. Here, we tried to investigate the effectiveness of adipose-derived mesenchymal stem cells (AD-MSC) on long-term lesions induced by CEES, an SM analog. The modulation of pulmonary immune cells and alveolar macrophage phenotype alteration was studied in the animal model used. Histopathological changes were investigated in the lungs and analysis of surface markers of alveolar macrophages as well as their cytokine expression in the BAL fluid was carried out by flow cytometry and ELISA, respectively. Treatment of mice with AD-MSC after intraperitoneal administration of CEES (10 mg/kg) reduces progressive histopathologic changes in the lung. Flow cytometric analysis of isolated alveolar macrophages in the bronchoalveolar lavage showed that the accumulation of both M1 and M2 macrophages in response to CEES was reduced by MSC administration. AD-MSCs caused a marked reduction in the CD86- and CD206-expressing macrophages compared to the untreated groups. The modulating effect of AD-MSCs in the M1-subset was much more significant compared to M2. These findings suggest that AD-MSCs understand their environment and restore the balance in disorders associated with Th1 or Th2 imbalance. Our results indicate that MSCs may represent an effective approach to repair lung injury induced by mustards.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Chlorethylethylsulfid, 97%