Merck
  • Suppressive activity of Vδ2+ γδ T cells on αβ T cells is licensed by TCR signaling and correlates with signal strength.
Suppressive activity of Vδ2+ γδ T cells on αβ T cells is licensed by TCR signaling and correlates with signal strength.
Cancer immunology, immunotherapy : CII (2020-01-27)
Karin Schilbach, Naomi Krickeberg, Carlotta Kaißer, Simon Mingram, Janika Kind, Gabrielle M Siegers, Hisayoshi Hashimoto
ZUSAMMENFASSUNG
Despite recent progress in the understanding of γδ T cells' roles and functions, their interaction with αβ T cells still remains to be elucidated. In this study, we sought to clarify what precisely endows peripheral Vδ2+ T cells with immunosuppressive function on autologous αβ T cells. We found that negatively freshly isolated Vδ2+ T cells do not exhibit suppressive behavior, even after stimulation with IL-12/IL-18/IL-15 or the sheer contact with butyrophilin-3A1-expressing tumor cell lines (U251 or SK-Mel-28). On the other hand, Vδ2+ T cells positively isolated through TCR crosslinking or after prolonged stimulation with isopentenyl pyrophosphate (IPP) mediate strong inhibitory effects on αβ T cell proliferation. Stimulation with IPP in the presence of IL-15 induces the most robust suppressive phenotype of Vδ2+ T cells. This indicates that Vδ2+ T cells' suppressive activity is dependent on a TCR signal and that the degree of suppression correlates with its strength. Vδ2+ T cell immunosuppression does not correlate with their Foxp3 expression but rather with their PD-L1 protein expression, evidenced by the massive reduction of suppressive activity when using a blocking antibody. In conclusion, pharmacologic stimulation of Vδ2+ T cells via the Vδ2 TCR for activation and expansion induces Vδ2+ T cells' potent killer activity while simultaneously licensing them to suppress αβ T cell responses. Taken together, the study is a further step to understand-in more detail-the suppressive activity of Vδ2+ γδ T cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isopentenylpyrophosphat Triammoniumsalz -Lösung, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)