Merck
PIAS1 is not suitable as a urothelial carcinoma biomarker protein and pharmacological target.
PloS one (2019-10-23)
Holger Hans Hermann Erb, Marlies Ebert, Ronja Kuhn, Lukas Donix, Axel Haferkamp, Robert Ian Seed, Eva Jüngel
ZUSAMMENFASSUNG
Urothelial cancer (UC) is one of the most common cancers in Europe and is also one of the costliest to treat. When first line therapies show initial success, around 50% of cancers relapse and proceed to metastasis. In this study we assessed the Protein inhibitor of activated signal transducers and activators of transcription (PIAS)1 as a potential therapeutic target in urothelial cancer. PIAS1 is a key regulator of STAT1 signalling and may be implicated in carcinogenesis. In contrast to other cancer types PIAS1 protein expression is not significantly different in malignant areas of UC specimens compared to non-malignant tissue. In addition, we found that down-regulation and overexpression of PIAS1 had no effect on the viability or colony forming ability of tested cell lines. Whilst other studies of PIAS1 suggest an important biological role in cancer, this study shows that PIAS1 has no influence on reducing the cytotoxic effects of Cisplatin or cell recovery after DNA damage induced by irradiation. Taken together, these in vitro data demonstrate that PIAS1 is not a promising therapeutic target in UC cancer as previously shown in different entities such as prostate cancer (PCa).