Merck
  • Shift of bacterial community and denitrification functional genes in biofilm electrode reactor in response to high salinity.
Shift of bacterial community and denitrification functional genes in biofilm electrode reactor in response to high salinity.
Environmental research (2020-02-23)
Siyuan Zhai, Min Ji, Yingxin Zhao, Xiao Su
ZUSAMMENFASSUNG
High salinity suppresses denitrification by inhibiting microorganism activities. The shift of microbial community and denitrification functional genes under salinity gradient was systematically investigated in a biofilm electrode reactor (BER) and biofilm reactor (BR) systems. Denitrification efficiency of both BER and BR was not significantly inhibited during the period of low salinity (0-2.0%). As the salinity increased to 2.5%, BER could overcome the impact of high salinity and maintained a relatively stable denitrification performance, and the effluent NO3--N was lower than 1.5 mg/L. High salinity (>2.5%) impoverished microbial diversity and altered the microbial community in both BER and BR. However, two genera Methylophaga and Methyloexplanations were enriched in BER due to electrochemical stimulation, which can tolerate high salinity (>3.0%). The relative abundance of Methylophaga in BER was almost 10 times as much as in BR. Paracoccus is a hydrogen autotrophic denitrifier, which was obviously inhibited with 1.0% NaCl. The hetertrophic denitrifiers were primarily responsible for the nitrate removal in the BER compared to the autotrophic denitrifiers. The abundance and proportion of denitrifying functional genes confirmed that main denitrifiers shift to salt-tolerant species (nirK-type denitrifiers) to reduce the toxic effects. The napA (2.2 × 108 to 6.5 × 108 copies/g biofilm) and nosZ (2.2 × 107 to 4.4 × 107 copies/g biofilm) genes were more abundant in BER compared to BR's, which was attributed to the enrichment of Methylophaga alcalica and Methyloversatilis universalis FAM5 in the BER. The results proved that BER had greater denitrification potential under high salinity (>2.0%) stress at the molecular level.