Merck
  • The Wnt/β-catenin/VASP positive feedback loop drives cell proliferation and migration in breast cancer.
The Wnt/β-catenin/VASP positive feedback loop drives cell proliferation and migration in breast cancer.
Oncogene (2019-12-14)
Kai Li, Jingwei Zhang, Yihao Tian, Yanqi He, Xiaolong Xu, Wenting Pan, Yang Gao, Fangfang Chen, Lei Wei
ZUSAMMENFASSUNG
Previous studies have shown that the main function of VASP is to regulate the cytoskeleton and play an important role in promoting tumor cell metastasis. In this study, we first reveal that VASP is located in the nucleus of breast cancer cells and elucidate a Wnt/β-catenin/VASP positive feedback loop. We identify that VASP is a target gene of Wnt/β-catenin signaling pathway, and activation of Wnt/β-catenin signaling pathway can significantly upregulate VASP protein expression, while upregulated VASP protein can in turn promote translocation of β-catenin and DVL3 proteins into the nucleus. In the nucleus, VASP, DVL3, β-catenin, and TCF4 can form VASP/DVL3/β-catenin/TCF4 protein complex, activating Wnt/β-catenin signaling pathway, and promoting the expression of target genes VASP, c-myc, and cyclin D1. Thus, our study reveals that there is a Wnt/β-catenin/VASP malignant positive feedback loop in breast cancer, which promotes the proliferation and migration of breast cancer cells, and breaking this positive feedback loop may provide new strategy for breast cancer treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Insulin -Lösung human, sterile-filtered, BioXtra, suitable for cell culture