Merck
  • Insight into the Cooperation of P-glycoprotein (ABCB1) and Breast Cancer Resistance Protein (ABCG2) at the Blood-Brain Barrier: A Case Study Examining Sorafenib Efflux Clearance.
Insight into the Cooperation of P-glycoprotein (ABCB1) and Breast Cancer Resistance Protein (ABCG2) at the Blood-Brain Barrier: A Case Study Examining Sorafenib Efflux Clearance.
Molecular Pharmacology (2012)
Agarwal S, Elmquist WF.