Merck
Strained, Photoejecting Ru(II) Complexes that are Cytotoxic Under Hypoxic Conditions.
Photochemistry and photobiology (2019-11-07)
John Roque, Dmytro Havrylyuk, Patrick C Barrett, Tariq Sainuddin, Julia McCain, Katsuya Colón, William T Sparks, Evan Bradner, Susan Monro, David Heidary, Colin G Cameron, Edith C Glazer, Sherri A McFarland
ZUSAMMENFASSUNG
A series of strained Ru(II) complexes were studied for potential anticancer activity in hypoxic tissues. The complexes were constructed with methylated ligands that were photolabile and an imidizo[4,5-f][1,10]phenanthroline ligand that contained an appended aromatic group to potentially allow for contributions of ligand-centered excited states. A systematic variation of the size and energy of the aromatic group was performed using systems containing 1-4 fused rings, and the photochemical and photobiological behaviors of all complexes were assessed. The structure and nature of the aromatic group had a subtle impact on photochemistry, altering environmental sensitivity, and had a significant impact on cellular cytotoxicity and photobiology. Up to 5-fold differences in cytotoxicity were observed in the absence of light activation; this rose to 50-fold differences upon exposure to 453 nm light. Most significantly, one complex retained activity under conditions with 1% O2 , which is used to induce hypoxic changes. This system exhibited a photocytotoxicity index (PI) of 15, which is in marked contrast to most other Ru(II) complexes, including those designed for O2 -independent mechanisms of action.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Naphthaldehyd, 95%
Sigma-Aldrich
Ruthenium(III)-chlorid, Anhydrous, solid, Ru content 45-55%
Sigma-Aldrich
1,10-Phenanthrolin-5,6-dion, 97%
Sigma-Aldrich
1-Pyrencarboxaldehyd, 99%