Merck
  • Development of a simple and rapid LC-MS/MS method for the simultaneous quantification of five Alternaria mycotoxins in human urine.
Development of a simple and rapid LC-MS/MS method for the simultaneous quantification of five Alternaria mycotoxins in human urine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2020-04-07)
Xiaoting Qiao, Jing Zhang, Yunjia Yang, Jie Yin, Hong Li, Yang Xing, Bing Shao
ZUSAMMENFASSUNG
Alternaria mycotoxins, such as tenuazonic acid (TeA), altenuene (ALT), alternariol (AOH), tentoxin (TEN) and alternariol monomethyl ether (AME) are frequently found in foods and may pose a potential risk to human health. Human biomonitoring can help measure our exposure to these mycotoxins, and help us determine if the exposure is changing over time. In this study, a simple liquid-liquid extraction sample preparation procedure followed by liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS) was developed for the simultaneous analysis of five Alternaria mycotoxins in human urine. High recoveries (92.7-103.2%) were obtained for all the tested mycotoxins with relative standard deviations (RSDs, %) of less than 6.4%. The limits of quantification (LOQs) for the analytes in urine ranged from 0.001 to 0.05 ng/mL. The method was successfully applied to investigate the levels of five Alternaria mycotoxins from 135 volunteers. In all of the samples, at least one Alternaria mycotoxin was detected. TeA, AME and AOH were the predominant Alternaria mycotoxins, and the detection rates were 85.9%, 96.3% and 51.9%, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Tenuazonsäure, analytical standard