Merck
  • Mechanism of deoxynivalenol-induced neurotoxicity in weaned piglets is linked to lipid peroxidation, dampened neurotransmitter levels, and interference with calcium signaling.
Mechanism of deoxynivalenol-induced neurotoxicity in weaned piglets is linked to lipid peroxidation, dampened neurotransmitter levels, and interference with calcium signaling.
Ecotoxicology and environmental safety (2020-03-09)
Xichun Wang, Xiaofang Chen, Li Cao, Lei Zhu, Yafei Zhang, Xiaoyan Chu, Dianfeng Zhu, Sajid Ur Rahman, Chenglu Peng, Shibin Feng, Yu Li, Jinjie Wu
ZUSAMMENFASSUNG
Deoxynivalenol(DON) has broad toxicity in livestock, but we know little about its neurotoxic mechanisms. We investigated DON neurotoxicity in the cerebral cortex, cerebellum, and hippocampus of "Duroc × Landrace × Yokshire" piglets. Control piglets were fed a basal diet, while those in low- and high-treatment groups were fed diets with 1.3 mg/kg and 2.2 mg/kg DON, respectively. After a 60 d trial, scanning electron microscopy revealed the destruction of hippocampal cell ultrastructure. As DON concentrations increased, oxidative damage also increased in the cerebral cortex, cerebellum, and hippocampus. Norepinephrine and 5-hydroxytryptamine concentrations tended to increase, whereas dopamine and γ-aminobutyric acid concentrations decreased. We also observed an increase in calcium concentration, relative mRNA expression of calcium/calmodulin-dependent protein kinase II (CaMKII), and CaMKII phosphorylation. However, calmodulin (CaM) mRNA and protein content decreased. Overall, our results suggest that DON acts through the Ca2+/CaM/CaMKII signaling pathway to influence cerebral lipid peroxidation and neurotransmitter levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deoxynivalenol
Sigma-Aldrich
6-Diazo-5-oxo-L-Norleucin, powder