Merck
  • Development and characterization of a human cell line-based transactivation assay to assess thyroid EDCs.
Development and characterization of a human cell line-based transactivation assay to assess thyroid EDCs.
Environmental research (2020-01-10)
Seok-Hee Lee, Hyeyeong Seo, Hee-Seok Lee, Yooheon Park
ZUSAMMENFASSUNG
Thyroid hormones (THs) are one of the most important hormones, playing key roles in the regulation of various physiological functions. Although THs have important function in human, in vitro test methods based on human cells are currently insufficient to effectively screen and test TH-related endocrine disrupting chemicals (EDCs). We established a TH agonist TA assay using the adenocarcinomic human alveolar basal epithelial cell line A549 to test and screen potential TH agonists. To establish the TH agonist TA assay, a TRE-secNluc-IRES-EGFP reporter cassette was constructed and transfected into the A549 cell line using a retrovirus. We evaluated the TH agonistic properties of several chemicals which were tested by existing thyroid agonists testing method (OECD GD 207). Comparing the results of the TH agonist TA assay with the OECD GD 207, T3, T4, tiratricol, and tetrac (natural TH and 3,3',5,5'-tetraiodothyroacetic acid derivatives), which are TH agonists according to the OECD GD 207, also tested positive in the TH agonist TA assay using the A549 cell line. These results suggested that the TH agonist TA assay developed in this study using a human cell line can provide the information, such as accuracy and specificity to TH agonistic properties of chemicals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2′,4,4′-Tetrahydroxybenzophenon, 97%
Sigma-Aldrich
3,3′,5,5′-Tetraiodthyroessigsäure
Sigma-Aldrich
3,3′,5-Triiodothyroessigsäure, ≥90%