Merck
  • Sphingosine 1-phosphate-regulated transcriptomes in heterogenous arterial and lymphatic endothelium of the aorta.
Sphingosine 1-phosphate-regulated transcriptomes in heterogenous arterial and lymphatic endothelium of the aorta.
eLife (2020-02-25)
Eric Engelbrecht, Michel V Levesque, Liqun He, Michael Vanlandewijck, Anja Nitzsche, Hira Niazi, Andrew Kuo, Sasha A Singh, Masanori Aikawa, Kristina Holton, Richard L Proia, Mari Kono, William T Pu, Eric Camerer, Christer Betsholtz, Timothy Hla
ZUSAMMENFASSUNG
Despite the medical importance of G protein-coupled receptors (GPCRs), in vivo cellular heterogeneity of GPCR signaling and downstream transcriptional responses are not understood. We report the comprehensive characterization of transcriptomes (bulk and single-cell) and chromatin domains regulated by sphingosine 1-phosphate receptor-1 (S1PR1) in adult mouse aortic endothelial cells. First, S1PR1 regulates NFκB and nuclear glucocorticoid receptor pathways to suppress inflammation-related mRNAs. Second, S1PR1 signaling in the heterogenous endothelial cell (EC) subtypes occurs at spatially-distinct areas of the aorta. For example, a transcriptomically distinct arterial EC population at vascular branch points (aEC1) exhibits ligand-independent S1PR1/ß-arrestin coupling. In contrast, circulatory S1P-dependent S1PR1/ß-arrestin coupling was observed in non-branch point aEC2 cells that exhibit an inflammatory gene expression signature. Moreover, S1P/S1PR1 signaling regulates the expression of lymphangiogenic and inflammation-related transcripts in an adventitial lymphatic EC (LEC) population in a ligand-dependent manner. These insights add resolution to existing concepts of endothelial heterogeneity, GPCR signaling and S1P biology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Esel-Serum
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Evans-Blau, Dye content ≥75 %
Sigma-Aldrich
Monoklonaler Anti-Aktin, α-Glattmuskel - Cy3 Maus-Antikörper, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Ausgeglichene Salzlösung nach Hanks 10x, Without sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-VCAM-1-Antikörper, Klon M/K, clone M/K - 2, Chemicon®, from rat