Merck
Transcription factor AP2 controls cnidarian germ cell induction.
Science (New York, N.Y.) (2020-02-15)
Timothy Q DuBuc, Christine E Schnitzler, Eleni Chrysostomou, Emma T McMahon, Febrimarsa, James M Gahan, Tara Buggie, Sebastian G Gornik, Shirley Hanley, Sofia N Barreira, Paul Gonzalez, Andreas D Baxevanis, Uri Frank
ZUSAMMENFASSUNG
Clonal animals do not sequester a germ line during embryogenesis. Instead, they have adult stem cells that contribute to somatic tissues or gametes. How germ fate is induced in these animals, and whether this process is related to bilaterian embryonic germline induction, is unknown. We show that transcription factor AP2 (Tfap2), a regulator of mammalian germ lines, acts to commit adult stem cells, known as i-cells, to the germ cell fate in the clonal cnidarian Hydractinia symbiolongicarpusTfap2 mutants lacked germ cells and gonads. Transplanted wild-type cells rescued gonad development but not germ cell induction in Tfap2 mutants. Forced expression of Tfap2 in i-cells converted them to germ cells. Therefore, Tfap2 is a regulator of germ cell commitment across germ line-sequestering and germ line-nonsequestering animals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
bisBenzimid H 33342, suitable for fluorescence, ≥97.0% (HPLC)