Merck
Stability of Mycotoxins in Individual Stock and Multi-Analyte Standard Solutions.
Toxins (2020-02-06)
Mariya Kiseleva, Zakhar Chalyy, Irina Sedova, Ilya Aksenov
ZUSAMMENFASSUNG
Standard solutions of mycotoxins prepared in RP HPLC solvents from neat standards are usually used for analytical method development. Multi-mycotoxin HPLC-MS/MS methods necessitate stability estimation for the wide spectrum of fungal metabolites. The stability of individual diluted stock standard solutions of mycotoxins in RP-HPLC solvents and multi-analyte HPLC-MS/MS calibrants was evaluated under standard storage and analysis conditions. Individual stock standard solutions of aflatoxins, sterigmatocystin, A- and B-trichothecenes, zearalenone and its analogues, ochratoxin A, fumonisins, Alternaria toxins, enniatins and beauvericin, moniliformin, citrinin, mycophenolic, cyclopiazonic acids and citreoviridin were prepared in RP-HPLC solvents and stored at -18 °C for 14 months. UV-spectroscopy was utilized to monitor the stability of analytes, excluding fumonisins. The gradual degradation of α-, β-zearalenol and α-, β-zearalanol in acetonitrile was detected. Aflatoxins and sterigmatocystin, zearalenone, Alternaria toxins, enniatins and beauvericin, citrinin, mycophenolic, cyclopiazonic acids and citreoviridin can be referred to as stable. The concentration of the majority of trichothecenes should be monitored. Diluted multi-mycotoxin standard in water/methanol (50/50 v/v) solutions acidified with 0.1% formic acid proved to be stable in silanized glass at 23 °C exposed to light for at least 75 h (CV≤10%). An unexpected manifestation of MS/MS signal suppression/enhancement was discovered in the course of multi-mycotoxin standard solution stability evaluation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Diazo-5-oxo-L-Norleucin, powder
Sigma-Aldrich
Fumonisin B1 aus Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
HT-2-Toxin, ≥98% (HPLC)
Supelco
Diacetoxyscirpenol -Lösung, ~100 μg/mL in acetonitrile, analytical standard