Merck
  • Monitoring the levels of brominated and organophosphate flame retardants in passenger cars: Utilisation of car air filters as active samplers.
Monitoring the levels of brominated and organophosphate flame retardants in passenger cars: Utilisation of car air filters as active samplers.
Journal of environmental sciences (China) (2020-03-17)
Long K Tran, Chang He, Dam Hoang Phuc, Leisa-Maree L Toms, Xianyu Wang, Meng Xiu, Jochen F Mueller, Adrian Covaci, Lidia Morawska, Phong K Thai
ZUSAMMENFASSUNG
Filters in residential and office air conditioning (A/C) systems have been used as sampling devices for monitoring different pollutants. However, cabin air filters (CAFs) in the A/C system of passenger cars have not been utilised for this purpose. In this study, we collected 22 used CAFs from passenger cars in Hanoi, Vietnam to analyse for 8 polybrominated diphenyl ethers (PBDEs) and 10 organophosphate esters (OPEs). All the analytes were detected in more than 50% of samples with the exception of BDE153 and BDE154. The average concentrations of ∑10OPEs and ∑8BDEs in the captured dust were 2600 and 40 ng/g, respectively with Tris (1-chloro-2-propyl) phosphate (TCIPP) and BDE209 as the dominant congener in OPE and BDE groups, respectively. CAFs are a potential tool to qualitatively assess the levels of semi-volatile chemicals in suspended dust in cars as a screening step for exposure assessment of those chemicals.