Merck
  • Multi-exposures to suspected endocrine disruptors in electronic waste recycling workers: Associations with thyroid and reproductive hormones.
Multi-exposures to suspected endocrine disruptors in electronic waste recycling workers: Associations with thyroid and reproductive hormones.
International journal of hygiene and environmental health (2020-01-22)
Sabrina Gravel, Jérôme Lavoué, Bouchra Bakhiyi, Jacques Lavoie, Brigitte Roberge, Louis Patry, Maryse F Bouchard, Marc-André Verner, Joseph Zayed, France Labrèche
ZUSAMMENFASSUNG
Electronic waste recycling (e-recycling) exposes workers to substances such as flame retardants and metals. Some of them are known or suspected endocrine disruptors that could affect hormonal homeostasis and eventually result in adverse health outcomes. Our aim was to measure biological concentrations of organophosphate ester (OPE) metabolites, polybrominated diphenyl ethers (PBDEs), mercury, lead and cadmium in e-recycling workers, and to explore associations with thyroid and sexual hormones. In a cross-sectional study, end-of-shift blood and urine spot samples were collected from 23 women and 77 men in six e-recycling facilities and one commercial recycling facility. Urinary concentrations of 15 OPE metabolites and mercury, and blood concentrations of 12 PBDE congeners, lead, cadmium, and thyroid (thyroxine [T4], triiodothyronine [T3], thyroid stimulating hormone [TSH]) and sexual (testosterone [T], estradiol, Follicle Stimulating Hormone [FSH], Luteinizing hormone [LH]) hormones were measured. E-recycling workers had higher concentrations of BDE209, all OPE metabolites, and lead than commercial recycling workers. In e-recycling workers, plasma geometric mean concentration of BDE209 was 18 ng/g lipids (geometric standard deviation [GSD]: 2.8) vs.1.7 ng/g lipids (GSD: 2.8) in commercial recycling, and urinary geometric mean concentration of diphenyl phosphate (DPhP), a major metabolite of triphenyl phosphate, was 1.7 ng/ml (GSD: 2.5), vs. 0.95 ng/ml (GSD: 2.0). In men, a two-fold increase in BDE209 was associated with 3.1% (95% Confidence interval: 0.07, 6.1) higher levels of total T4, and a two-fold increase in tert-butyl diphenyl phosphate (tb-DPhP) was associated with 18% (-29, -4.7) lower total T, 18% (-27, -6.9) lower free T and 13% (-25, 0.70) lower free T/estradiol ratio. In women, a two-fold increase in BDE153 was associated with 10% (-17, -3.2) lower free T3. To our knowledge, this is the first study to show associations between OPE metabolites and sex hormones in adults. Although some of our results are not conclusive and need replication, they suggest that prudent avoidance should be applied in risk management of flame retardants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Supelco
TDCPP, PESTANAL®, analytical standard